Assay Details
Target Gene Details
Entrez Gene ID: | 241769 |
Gene Name: | potassium channel, subfamily K, member 15 |
Gene Aliases: |
KCNK11, KCNK14, KT3.3, TASK5 |
Location: |
Chr.2:163853750-163858874 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnk15 | NM_001030292.1 | 1 | 56 | NP_001025463.1 |
| BC147255.1 | 1 | 155 | ||
| BC147256.1 | 1 | 155 |
More Information
Set Membership: |
|

Genomic Map