Assay Details
Target Gene Details
Entrez Gene ID: | 27054 |
Gene Name: | SEC23B (S. cerevisiae) |
Gene Aliases: |
- |
Location: |
Chr.2:144556229-144590753 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sec23b | NM_001252543.1 | 20 | 2407 | NP_001239472.1 |
| NM_001252544.1 | 20 | 2400 | NP_001239473.1 | |
| NM_001252545.1 | 20 | 2389 | NP_001239474.1 | |
| NM_001281816.1 | 17 | 2029 | NP_001268745.1 | |
| NM_019787.4 | 20 | 2418 | NP_062761.2 | |
| AF200326.1 | AAF08301.1 | |||
| AK014597.1 | 20 | 2368 | BAB29452.1 | |
| AK029661.1 | 17 | 2030 | BAC26553.1 | |
| AK139729.1 | 20 | 2333 | ||
| AK146083.1 | 20 | 2367 | BAE26887.1 | |
| AK149693.1 | 20 | 2347 | BAE29030.1 | |
| AK153214.1 | 20 | 2374 | BAE31812.1 | |
| AK155168.1 | BAE33090.1 | |||
| AK158398.1 | 20 | 2385 | BAE34488.1 | |
| AK166330.1 | 20 | 2350 | BAE38710.1 | |
| AK167382.1 | 20 | 2355 | ||
| AK167732.1 | 20 | 2336 | BAE39773.1 | |
| AK171597.1 | 20 | 2348 | BAE42552.1 | |
| BC005464.1 | 20 | 2343 | AAH05464.1 | |
| BC011160.1 | 20 | 2370 | AAH11160.1 | |
| BP766886.1 | 1 | 436 | ||
| CK619007.1 | 1 | 396 | ||
| CK623414.1 |
More Information
Set Membership: |
|

Genomic Map