Assay Details
Target Gene Details
Entrez Gene ID: | 76688 |
Gene Name: | ADP-ribosylation factor related protein 1 |
Gene Aliases: |
1500006I01Rik, AI480700 |
Location: |
Chr.2:181357690-181365404 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arfrp1 | NM_001165991.1 | 8 | 1011 | NP_001159463.1 |
| NM_001165992.1 | 8 | 992 | NP_001159464.1 | |
| NM_001165995.1 | 7 | 898 | NP_001159467.1 | |
| NM_029702.4 | 8 | 1005 | NP_083978.3 | |
| NR_028585.1 | 7 | 934 | ||
| AJ413953.1 | 8 | 958 | CAC88692.1 | |
| AK032592.1 | 2 | 789 | ||
| AK044759.1 | 8 | 963 | BAC32069.1 | |
| AK050136.1 | ||||
| AK184494.1 | 1 | 229 | ||
| BC010713.1 | AAH10713.1 | |||
| BC046782.1 | 7 | 872 | AAH46782.1 | |
| BC046786.1 | 7 | 872 | AAH46786.1 |

Genomic Map