Assay Details
Target Gene Details
Entrez Gene ID: | 211484 |
Gene Name: | testis specific 10 |
Gene Aliases: |
4933432N21Rik, AI843383, Gm217, Mtsga10 |
Location: |
Chr.1:37728144-37866739 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tsga10 | NM_207228.3 | 20 | 2912 | NP_997111.1 |
| AK081029.1 | 1 | 174 | ||
| BC066782.1 | 20 | 2909 | AAH66782.1 |
More Information
Set Membership: |
|

Genomic Map