Assay Details
Target Gene Details
Entrez Gene ID: | 20872 |
Gene Name: | serine/threonine kinase 16 |
Gene Aliases: |
EDPK, Krct, MPSK, PKL12, TSF-1 |
Location: |
Chr.1:75210829-75215606 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stk16 | NM_001277992.1 | 8 | 2769 | NP_001264921.1 |
| NM_011494.5 | 8 | 2797 | NP_035624.3 | |
| NR_102731.1 | 8 | 2817 | ||
| AK030404.1 | 8 | 2772 | BAC26947.1 |
Target Gene Details
Entrez Gene ID: | 22145 |
Gene Name: | tubulin, alpha 4A |
Gene Aliases: |
M[a]4, Tuba4 |
Location: |
Chr.1:75214974-75219860 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tuba4a | NM_009447.3 | 4 | 1509 | NP_033473.1 |
| AK002427.1 | BAB22094.1 | |||
| AK078355.1 | 4 | 1510 | BAC37234.1 | |
| AK158934.1 | 4 | 1538 | BAE34732.1 | |
| BC019959.1 | 4 | 1487 | AAH19959.1 |
More Information
Set Membership: |
|

Genomic Map