Assay Details
Target Gene Details
Entrez Gene ID: | 13345 |
Gene Name: | twist basic helix-loop-helix transcription factor 2 |
Gene Aliases: |
Dermo1, bHLHa39 |
Location: |
Chr.1:91801477-91848027 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Twist2 | NM_007855.2 | 2 | 884 | NP_031881.1 |
| AK008388.1 | 2 | 622 | ||
| AK008411.1 | 2 | 621 | ||
| AK011180.1 | 2 | 900 | BAB27450.1 | |
| AK011861.1 | 2 | 901 | BAB27885.1 | |
| BC090636.1 | 2 | 889 | ||
| U36384.1 | 2 | 884 | AAC52319.1 |
More Information
Set Membership: |
|

Genomic Map