Assay Details
Target Gene Details
Entrez Gene ID: | 70933 |
Gene Name: | RIKEN cDNA 4931402G19 gene |
Gene Aliases: |
- |
Location: |
Chr.2:120455698-120469367 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4931402G19Rik | NR_040608.1 | |||
| AK016423.1 |
Target Gene Details
Entrez Gene ID: | 12335 |
Gene Name: | calpain 3 |
Gene Aliases: |
AI323605, Capa-3, Capa3, Lp82, p94 |
Location: |
Chr.2:120463593-120504919 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Capn3 | NM_001177799.1 | 1 | 390 | NP_001171270.1 |
| NM_007601.3 | 1 | 390 | NP_031627.2 | |
| AF127766.2 | 1 | 110 | AAD28255.2 | |
| BC107011.2 | 1 | 110 | ||
| BC139790.1 | 1 | 110 | ||
| CN664345.1 | 1 | 370 | ||
| X92523.1 | 1 | 110 | CAA63301.1 |
More Information
Set Membership: |
|

Genomic Map