Assay Details
Target Gene Details
Entrez Gene ID: | 14683 |
Gene Name: | GNAS (guanine nucleotide binding protein, alpha stimulating) complex locus |
Gene Aliases: |
5530400H20Rik, A930027G11Rik, C130027O20Rik, GPSA, GSP, Galphas, Gnas1, Gnasxl, Gsa, Nesp, Nespl, Oed-Sml, Oedsml, P1, P2, P3, PHP1A, PHP1B, POH |
Location: |
Chr.2:174283444-174346744 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gnas | NM_001077507.1 | NP_001070975.1 | ||
| NM_001077510.2 | NP_001070978.1 | |||
| NM_010309.3 | NP_034439.2 | |||
| NM_019690.2 | 5 | 1567 | NP_062664.2 | |
| NM_022000.2 | NP_068840.2 | |||
| NM_201616.1 | NP_963910.1 | |||
| NM_201618.1 | NP_963912.1 | |||
| NR_003258.1 | ||||
| AF107847.1 | 5 | 1571 | AAD11805.1 | |
| AF107848.1 | AAD11806.1 | |||
| AF116268.1 | AAD14686.1 | |||
| AJ010163.1 | CAA09026.1 | |||
| AK078037.1 | ||||
| AK132636.1 | ||||
| AK147051.1 | BAE27636.1 | |||
| AK159563.1 | BAE35187.1 | |||
| AK168996.1 | BAE40795.1 | |||
| AK185862.1 | ||||
| AK197380.1 | ||||
| AY519501.1 | AAS00601.1 | |||
| AY519502.1 | AAS00602.1 | |||
| AY519504.1 | AAS00604.1 | |||
| AY682719.1 | ||||
| BC038067.1 | AAH38067.1 | |||
| BC061496.1 | AAH61496.1 | |||
| BC062654.1 | AAH62654.1 | |||
| BC080816.1 | AAH80816.1 | |||
| BC092055.1 | AAH92055.1 | |||
| BC106133.1 | AAI06134.1 | |||
| BF468141.1 | ||||
| M13964.1 | AAA37745.1 | |||
| S49979.1 | AAB24002.1 | |||
| S49980.1 | AAB24003.1 | |||
| S79606.1 | ||||
| Y00703.1 | CAA68695.1 |
More Information
Set Membership: |
|

Genomic Map