Assay Details
Target Gene Details
Entrez Gene ID: | 16558 |
Gene Name: | kinesin family member 16B |
Gene Aliases: |
8430434E15Rik, AA623607, AI789011, C80253, C80902, mKIAA1590 |
Location: |
Chr.2:142618334-142901595 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kif16b | NM_001081133.2 | 26 | 4656 | NP_001074602.1 |
| AB610968.1 | 27 | 4744 | ||
| AK018455.1 | 2 | 1126 | ||
| AK129401.1 | 14 | 2860 | BAC98211.1 | |
| AK142909.1 | 8 | 1957 | ||
| AK160835.1 | 10 | 2876 | BAE36039.1 |
More Information
Set Membership: |
|

Genomic Map