Assay Details
Target Gene Details
Entrez Gene ID: | 329504 |
Gene Name: | leucine carboxyl methyltransferase 2 |
Gene Aliases: |
D330024M17, Tyw4 |
Location: |
Chr.2:121137292-121140698 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lcmt2 | NM_177846.3 | 1 | 1709 | NP_808514.2 |
| AK054392.1 | 1 | 1399 | BAC35760.1 | |
| AK084642.1 | 1 | 1676 | ||
| AK172552.1 | 1 | 1390 | BAE43064.1 | |
| AK172970.1 | 1 | 1664 | BAD32248.1 | |
| AK178658.1 |
More Information
Set Membership: |
|

Genomic Map