Assay Details
Target Gene Details
Entrez Gene ID: | 73649 |
Gene Name: | cytochrome b reductase 1 |
Gene Aliases: |
2210407P13Rik, Dcytb |
Location: |
Chr.2:71118054-71142926 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cybrd1 | NM_028593.2 | 1 | 177 | NP_082869.2 |
| AF354666.1 | 1 | 119 | AAK50909.1 | |
| AK008849.1 | 1 | 123 | BAB25928.1 | |
| AK029112.1 | 1 | 180 | BAC26304.1 |
More Information
Set Membership: |
|

Genomic Map