Assay Details
Target Gene Details
Entrez Gene ID: | 68236 |
Gene Name: | gametocyte specific factor 1-like |
Gene Aliases: |
2410116G06Rik |
Location: |
Chr.2:163087031-163089601 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gtsf1l | NM_026630.2 | 2 | 204 | NP_080906.1 |
AK018886.1 | 1 | 144 | BAB31474.1 | |
AK019131.1 | 2 | 205 | BAB31558.1 | |
AK160150.1 | 1 | 104 | BAE35658.1 | |
BC048439.1 | 1 | 77 | AAH48439.1 |
