Assay Details
Target Gene Details
Entrez Gene ID: | 99011 |
Gene Name: | protein-O-mannosyltransferase 1 |
Gene Aliases: |
AI505244 |
Location: |
Chr.2:32236595-32255005 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pomt1 | NM_145145.1 | 20 | 2704 | NP_660127.1 |
| AK053889.1 | 19 | 3224 | BAC35577.1 | |
| AK134770.1 | 20 | 2748 | BAE22274.1 | |
| AK153984.1 | 20 | 2803 | BAE32295.1 | |
| AK202135.1 | 1 | 110 | ||
| AY007238.1 | 20 | 2744 | AAG15588.1 | |
| AY494857.1 | 20 | 2764 | AAS76201.1 | |
| BC027325.1 | 20 | 2704 | AAH27325.1 |
More Information
Set Membership: |
|

Genomic Map