Assay Details
Target Gene Details
Entrez Gene ID: | 13636 |
Gene Name: | ephrin A1 |
Gene Aliases: |
AI325262, B61, Efl1, Epl1, Eplg1, Lerk1 |
Location: |
Chr.3:89271730-89280951 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Efna1 | NM_001162425.1 | 5 | 728 | NP_001155897.1 |
| NM_010107.4 | 5 | 764 | NP_034237.3 | |
| AK009144.1 | 5 | 761 | BAB26102.1 | |
| AK079266.1 | 5 | 731 | ||
| BC002046.1 | 5 | 729 | AAH02046.1 | |
| D38146.1 | BAA07344.1 | |||
| U26188.1 | 5 | 723 | AAA67563.1 | |
| U90662.1 | 5 | 663 | AAB50237.1 |
More Information
Set Membership: |
|

Genomic Map