Assay Details
Target Gene Details
Entrez Gene ID: | 17116 |
Gene Name: | mab-21-like 1 (C. elegans) |
Gene Aliases: |
AW047968 |
Location: |
Chr.3:55782510-55785287 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mab21l1 | NM_010750.3 | 1 | 1860 | NP_034880.1 |
| AF228913.1 | 1 | 1845 | AAF67140.1 | |
| AK032114.1 | 1 | 1831 | BAC27711.1 | |
| AK043552.1 | 1 | 1859 | ||
| BC014750.1 | 1 | 1809 | AAH14750.1 |
Target Gene Details
Entrez Gene ID: | 26422 |
Gene Name: | neurobeachin |
Gene Aliases: |
Lyst2, mKIAA1544 |
Location: |
Chr.3:55625195-56183701 on Build GRCm38 |
Assay Gene Location: | Within Intron 41 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nbea | NM_030595.1 | NP_085098.1 | ||
| AK043125.1 | BAC31466.1 | |||
| Y18276.1 | CAC18811.1 |
More Information
Set Membership: |
|

Genomic Map