Assay Details
Target Gene Details
Entrez Gene ID: | 66860 |
Gene Name: | tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 1 |
Gene Aliases: |
1200003E16Rik, Tanc, mKIAA1728 |
Location: |
Chr.2:59611848-59846213 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tanc1 | NM_001290659.1 | 13 | 2203 | NP_001277588.1 |
| NM_198294.3 | 14 | 2379 | NP_938036.2 | |
| AB093299.1 | 3 | 472 | BAC41481.1 | |
| AK030022.1 | 11 | 1983 | BAC26741.1 | |
| BC079914.1 | 13 | 2182 | ||
| BC141367.1 | 13 | 2193 |

Genomic Map