Assay Details
Target Gene Details
Entrez Gene ID: | 170439 |
Gene Name: | ELOVL family member 6, elongation of long chain fatty acids (yeast) |
Gene Aliases: |
C77826, FAE, LCE |
Location: |
Chr.3:129532386-129638495 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elovl6 | NM_130450.2 | 5 | 4138 | NP_569717.1 |
| AB072039.1 | 6 | 4051 | BAB68544.1 | |
| AY053453.1 | 5 | 4119 | AAL14239.1 |
More Information
Set Membership: |
|

Genomic Map