Assay Details
Target Gene Details
Entrez Gene ID: | 433586 |
Gene Name: | mastermind like 3 (Drosophila) |
Gene Aliases: |
AV234550, BC049812, Mam-2, mKIAA1816 |
Location: |
Chr.3:51687320-52105085 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Maml3 | NM_001004176.2 | 3 | 3002 | NP_001004176.2 |
| AB553633.1 | 3 | 2568 | ||
| AK157983.1 | 2 | 234 | BAE34300.1 | |
| AK173269.1 | 2 | 1212 | BAD32547.1 | |
| BC085318.1 | 2 | 891 | AAH85318.1 |
More Information
Set Membership: |
|

Genomic Map