Assay Details
Target Gene Details
Entrez Gene ID: | 433586 |
Gene Name: | mastermind like 3 (Drosophila) |
Gene Aliases: |
AV234550, BC049812, Mam-2, mKIAA1816 |
Location: |
Chr.3:51687320-52105085 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Maml3 | NM_001004176.2 | 5 | 5434 | NP_001004176.2 |
| AB553633.1 | 5 | 5000 | ||
| AK144864.1 | 1 | 1005 | ||
| AK173269.1 | 4 | 3644 | BAD32547.1 | |
| BC049812.1 | 1 | 1934 | AAH49812.1 |
More Information
Set Membership: |
|

Genomic Map