Assay Details
Target Gene Details
Entrez Gene ID: | 54635 |
Gene Name: | platelet-derived growth factor, C polypeptide |
Gene Aliases: |
1110064L01Rik, AI647969, PDGF-C |
Location: |
Chr.3:81035885-81214040 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdgfc | NM_019971.2 | 1 | 945 | NP_064355.1 |
| AF117608.1 | 1 | 121 | AAF22516.1 | |
| AF266467.1 | 1 | 945 | AAK58566.1 | |
| AK033734.1 | 1 | 674 | BAC28455.1 | |
| AK042767.1 | 1 | 692 | BAC31358.1 | |
| AK052947.1 | 1 | 197 | BAC35216.1 | |
| BC037696.1 | 1 | 132 | AAH37696.1 |
More Information
Set Membership: |
|

Genomic Map