Assay Details
Target Gene Details
Entrez Gene ID: | 20198 |
Gene Name: | S100 calcium binding protein A4 |
Gene Aliases: |
18A2, 42a, Capl, FSp1, Mts1, PeL98, metastasin, pk9a |
Location: |
Chr.3:90603770-90606045 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100a4 | NM_011311.2 | 3 | 297 | NP_035441.1 |
| AK003065.1 | 3 | 303 | BAB22543.1 | |
| AK160522.1 | 3 | 298 | BAE35841.1 | |
| BC051214.1 | 2 | 256 | AAH51214.1 | |
| BC094635.1 | 2 | 256 | AAH94635.1 | |
| BX526220.1 | 2 | 248 | ||
| D00208.1 | 3 | 297 | BAA00148.1 | |
| X05835.1 | 2 | 254 | CAA29282.1 | |
| X16190.1 | 2 | 256 | CAA34316.1 |
More Information
Set Membership: |
|

Genomic Map