Assay Details
Target Gene Details
Entrez Gene ID: | 329735 |
Gene Name: | RIKEN cDNA 4933431E20 gene |
Gene Aliases: |
- |
Location: |
Chr.3:107888850-107896213 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4933431E20Rik | NR_015459.1 | 3 | 1516 | |
| AK030251.1 | 4 | 1139 | ||
| AK036227.1 | 3 | 1089 | BAC29352.1 | |
| AK147824.1 | 4 | 1113 | BAE28162.1 |
Target Gene Details
Entrez Gene ID: | 99662 |
Gene Name: | EPS8-like 3 |
Gene Aliases: |
AI504489, Eps8r3 |
Location: |
Chr.3:107873860-107892900 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eps8l3 | NM_133867.2 | 19 | 2111 | NP_598628.1 |
| AK136584.1 | 19 | 2108 | ||
| AY074932.1 | 19 | 2127 | AAL76121.1 | |
| BC014734.1 | 19 | 2099 | AAH14734.1 |
More Information
Set Membership: |
|

Genomic Map