Assay Details
Target Gene Details
Entrez Gene ID: | 14066 |
Gene Name: | coagulation factor III |
Gene Aliases: |
AA409063, CD142, Cf-3, Cf3, TF |
Location: |
Chr.3:121723537-121735052 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F3 | NM_010171.3 | 1 | 89 | NP_034301.3 |
| AK137642.1 | ||||
| AK144872.1 | ||||
| BC016397.1 | 1 | 80 | AAH16397.1 | |
| BC024886.1 | 1 | 89 | AAH24886.1 | |
| M26071.1 | AAA40414.1 |
More Information
Set Membership: |
|

Genomic Map