Assay Details
Target Gene Details
Entrez Gene ID: | 14013 |
Gene Name: | MDS1 and EVI1 complex locus |
Gene Aliases: |
D630039M04Rik, Evi-1, Evi1, Jbo, Mds, Mds1, Mds1-Evi1, Prdm3, Znfpr1b1 |
Location: |
Chr.3:29951296-30509783 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mecom | NM_021442.2 | 2 | 351 | NP_067417.1 |
| AJ010015.1 | 2 | 351 | CAA08971.1 |
More Information
Set Membership: |
|

Genomic Map