Assay Details
Target Gene Details
Entrez Gene ID: | 211550 |
Gene Name: | TRAF-interacting protein with forkhead-associated domain |
Gene Aliases: |
T2bp |
Location: |
Chr.3:127788857-127798394 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tifa | NM_145133.3 | 3 | 918 | NP_660115.1 |
| AB062111.1 | 3 | 715 | BAB86903.1 | |
| AK041891.1 | 2 | 752 | BAE43315.1 | |
| AK044221.1 | 2 | 862 | BAC31825.1 | |
| AK149408.1 | 3 | 754 | BAE28856.1 | |
| AK149864.1 | 3 | 886 | BAE29133.1 | |
| AK151651.1 | 3 | 869 | BAE30580.1 | |
| AK166239.1 | 3 | 960 | BAE38653.1 | |
| BC065775.1 | 3 | 913 | AAH65775.1 |

Genomic Map