Assay Details
Target Gene Details
Entrez Gene ID: | 228875 |
Gene Name: | solute carrier family 35, member C2 |
Gene Aliases: |
C85957, CGI-15, D2Wsu58e, Ovcov1 |
Location: |
Chr.2:165276522-165287888 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc35c2 | NM_001252573.1 | 10 | 1544 | NP_001239502.1 |
| NM_001252574.1 | 10 | 1479 | NP_001239503.1 | |
| NM_001252575.1 | 10 | 1427 | NP_001239504.1 | |
| NM_144893.2 | 10 | 1498 | NP_659142.1 | |
| NR_045544.1 | 11 | 1375 | ||
| AK053180.1 | 10 | 1414 | ||
| AK155539.1 | 10 | 1513 | BAE33316.1 | |
| AK160899.1 | 10 | 1484 | BAE36078.1 | |
| AK170715.1 | 10 | 1449 | BAE41973.1 | |
| AK195585.1 | ||||
| BB772752.1 | ||||
| BC018327.1 | 11 | 1364 | AAH18327.1 | |
| BC064092.1 | 2 | 631 | AAH64092.1 | |
| BC094025.1 | 10 | 1477 | AAH94025.1 | |
| BE952356.1 | 1 | 499 |
More Information
Set Membership: |
|

Genomic Map