Assay Details
Target Gene Details
Entrez Gene ID: | 329628 |
Gene Name: | FAT tumor suppressor homolog 4 (Drosophila) |
Gene Aliases: |
6030410K14Rik, 9430004M15 |
Location: |
Chr.3:38886487-39011987 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fat4 | NM_183221.3 | 4 | 5761 | NP_899044.3 |
| AK034552.1 | 4 | 1759 | BAC28751.1 | |
| DQ286572.1 | 4 | 5761 | ABB88946.1 |
More Information
Set Membership: |
|

Genomic Map