Assay Details
Target Gene Details
Entrez Gene ID: | 170439 |
Gene Name: | ELOVL family member 6, elongation of long chain fatty acids (yeast) |
Gene Aliases: |
C77826, FAE, LCE |
Location: |
Chr.3:129532386-129638495 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elovl6 | NM_130450.2 | NP_569717.1 | ||
| AB072039.1 | BAB68544.1 | |||
| AF480860.1 | AAM13450.1 | |||
| AK029029.1 | BAC26252.1 | |||
| AK087440.1 | ||||
| AK165847.1 | ||||
| AK167373.1 | BAE39469.1 | |||
| AY053453.1 | AAL14239.1 | |||
| BC051041.1 | AAH51041.1 | |||
| BC098492.1 | AAH98492.1 | |||
| BC100576.1 | AAI00577.1 |
More Information
Set Membership: |
|

Genomic Map