Assay Details
Target Gene Details
Entrez Gene ID: | 242050 |
Gene Name: | immunoglobulin superfamily, member 10 |
Gene Aliases: |
6530405F15Rik, 9030224D03, AA409708, AA536958, AI414626, Adlican2, CMF608 |
Location: |
Chr.3:59316735-59344764 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Igsf10 | NM_001162884.1 | 5 | 4827 | NP_001156356.1 |
| AK081990.1 | 1 | 372 |
More Information
Set Membership: |
|

Genomic Map