Assay Details
Target Gene Details
Entrez Gene ID: | 18741 |
Gene Name: | paired-like homeodomain transcription factor 2 |
Gene Aliases: |
9430085M16Rik, Brx1, Brx1b, Munc30, Otlx2, Ptx2, Rieg |
Location: |
Chr.3:129199878-129219594 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pitx2 | NM_001042502.2 | 1 | 429 | NP_001035967.1 |
| NM_001042504.2 | NP_001035969.1 | |||
| NM_001286942.1 | NP_001273871.1 | |||
| NM_001287048.1 | 1 | 429 | NP_001273977.1 | |
| NM_011098.4 | NP_035228.2 | |||
| AB006320.1 | BAA75247.1 | |||
| AB006321.1 | BAA75248.1 | |||
| AF048723.1 | AAC40086.1 | |||
| AF048724.1 | 1 | 389 | AAC40087.1 | |
| AJ243597.1 | 1 | 387 | CAB65259.1 | |
| AK034846.1 | 1 | 431 | ||
| AK035083.1 | 1 | 87 | BAC28939.1 | |
| AM940438.1 | ||||
| BB626986.1 | 1 | 431 | ||
| BC075660.1 | 1 | 94 | AAH75660.1 | |
| CJ177526.1 | ||||
| U80010.1 | AAC53119.1 | |||
| U80011.1 | AAC53120.1 | |||
| U80036.1 | AAB38505.1 |
More Information
Set Membership: |
|

Genomic Map