Assay Details
Target Gene Details
Entrez Gene ID: | 66209 |
Gene Name: | INTS3 and NABP interacting protein |
Gene Aliases: |
1110054O05Rik, 2610312O17Rik, AA399876, Ssbip1 |
Location: |
Chr.4:59769642-59783855 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Inip | NM_001013577.1 | 5 | 1810 | NP_001013595.1 |
| AK008870.1 | 1 | 407 | ||
| DQ691672.1 |

Genomic Map