Assay Details
Target Gene Details
Entrez Gene ID: | 68075 |
Gene Name: | leucine rich adaptor protein 1 |
Gene Aliases: |
1520402A15Rik, Lrp35a |
Location: |
Chr.4:116136668-116144616 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lurap1 | NM_026547.1 | 2 | 879 | NP_080823.1 |
| AK013564.1 | BAB28908.1 | |||
| BC147058.1 | 2 | 836 | ||
| BC147059.1 | 2 | 836 | ||
| BC147737.1 | 2 | 836 | ||
| BC147750.1 | 2 | 836 |

Genomic Map