Assay Details
Target Gene Details
Entrez Gene ID: | 66350 |
Gene Name: | phospholipase A2, group XIIA |
Gene Aliases: |
2310004B05Rik, GXII, GXII-1, GXII-1-PLA2, Pla2g12, Rossy, mGXII-1 |
Location: |
Chr.3:129878606-129895825 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pla2g12a | NM_001286948.1 | 1 | 228 | NP_001273877.1 |
| NM_023196.4 | 1 | 228 | NP_075685.2 | |
| NR_104611.1 | 1 | 228 | ||
| AK003183.1 | 1 | 187 | ||
| AK009133.1 | 1 | 230 | BAB26094.1 | |
| AK010011.1 | 1 | 218 | BAB26641.1 | |
| AK010174.1 | 1 | 194 | BAB26747.1 | |
| AK012414.1 | 1 | 157 | BAC25368.1 | |
| AK163693.1 | 1 | 142 | BAE37460.1 | |
| AY007381.1 | 1 | 201 | AAG23336.1 | |
| BC026812.1 | 1 | 193 | AAH26812.1 | |
| BC051117.1 | 1 | 228 | AAH51117.1 |
More Information
Set Membership: |
|

Genomic Map