Assay Details
Target Gene Details
Entrez Gene ID: | 654812 |
Gene Name: | angiopoietin-like 7 |
Gene Aliases: |
- |
Location: |
Chr.4:148492979-148500462 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Angptl7 | NM_001039554.3 | 1 | 355 | NP_001034643.1 |
| AK031675.1 | 1 | 355 | BAC27507.1 | |
| BC023373.1 | 1 | 342 | AAH23373.1 |
Target Gene Details
Entrez Gene ID: | 56717 |
Gene Name: | mechanistic target of rapamycin (serine/threonine kinase) |
Gene Aliases: |
2610315D21Rik, AI327068, FRAP, FRAP2, Frap1, RAFT1, RAPT1, flat |
Location: |
Chr.4:148448582-148557685 on Build GRCm38 |
Assay Gene Location: | Within Intron 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mtor | NM_020009.2 | NP_064393.2 | ||
| AF152838.1 | AAF73196.1 | |||
| AK143381.1 | ||||
| AK158715.1 | ||||
| BC112904.1 | AAI12905.1 |
More Information
Set Membership: |
|

Genomic Map