Assay Details
Target Gene Details
Entrez Gene ID: | 52837 |
Gene Name: | thioredoxin-related transmembrane protein 4 |
Gene Aliases: |
2810417D04Rik, 4930500L08Rik, AI843224, AW046784, D2Bwg1356e, Txndc13, mKIAA1162 |
Location: |
Chr.2:134594501-134644121 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmx4 | NM_029148.1 | 8 | 2962 | NP_083424.1 |
| AK013100.1 | 1 | 146 | ||
| AK129301.1 | 8 | 2961 | BAC98111.1 | |
| AK147465.1 | 8 | 2963 | BAE27931.1 | |
| BC050918.1 | 8 | 2861 | AAH50918.1 | |
| BC054558.1 | 8 | 2962 |

Genomic Map