Assay Details
Target Gene Details
Entrez Gene ID: | 22138 |
Gene Name: | titin |
Gene Aliases: |
1100001C23Rik, 2310036G12Rik, 2310057K23Rik, 2310074I15Rik, AF006999, AV006427, D330041I19Rik, D830007G01Rik, L56, mdm, shru |
Location: |
Chr.2:76703980-76982557 on Build GRCm38 |
Assay Gene Location: | Within Exon 313 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttn | NM_011652.3 | 312 | 100275 | NP_035782.3 |
| NM_028004.2 | 191 | 80532 | NP_082280.2 | |
| AK003152.1 | 3 | 690 | BAB22606.1 | |
| AK029024.1 | 5 | 1373 | BAC26249.1 | |
| BC025840.1 | AAH25840.1 |

Genomic Map