Assay Details
Target Gene Details
Entrez Gene ID: | 67037 |
Gene Name: | polyamine-modulated factor 1 |
Gene Aliases: |
2600009M07Rik, AL033286, AW060657 |
Location: |
Chr.3:88394138-88410408 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pmf1 | NM_025928.3 | 5 | 851 | NP_080204.1 |
| AF348509.1 | 3 | 654 | AAK84828.1 | |
| AF348510.1 | 5 | 861 | AAK84829.1 | |
| AK011174.1 | 5 | 853 | BAB27447.1 | |
| AK012113.1 | 5 | 852 | BAB28041.1 | |
| AK013787.1 | 5 | 854 | BAB28994.1 | |
| AK078308.1 | 5 | 871 | BAC37213.1 | |
| AK150603.1 | 5 | 868 | BAE29695.1 | |
| AK168200.1 | 5 | 852 | BAE40160.1 | |
| BC034333.1 | 5 | 829 | AAH34333.1 | |
| BC037139.1 | 5 | 860 | AAH37139.1 | |
| BC049223.1 | 5 | 850 | AAH49223.1 |

Genomic Map