Assay Details
Target Gene Details
Entrez Gene ID: | 18033 |
Gene Name: | nuclear factor of kappa light polypeptide gene enhancer in B cells 1, p105 |
Gene Aliases: |
NF-KB1, NF-kappaB, NF-kappaB1, p105, p50, p50/p105 |
Location: |
Chr.3:135584655-135691547 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nfkb1 | NM_008689.2 | 1 | 70 | NP_032715.2 |
| BC050841.1 | 1 | 70 | AAH50841.1 |
More Information
Set Membership: |
|

Genomic Map