Assay Details
Target Gene Details
Entrez Gene ID: | 64406 |
Gene Name: | trans-acting transcription factor 5 |
Gene Aliases: |
- |
Location: |
Chr.2:70474923-70477729 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sp5 | NM_022435.2 | 2 | 1528 | NP_071880.1 |
| AF279479.1 | 2 | 1285 | AAF87798.1 | |
| AK036234.1 | 2 | 1528 | BAC29355.1 | |
| BC116691.1 | 2 | 1379 | ||
| BC117505.1 | 2 | 1379 |
More Information
Set Membership: |
|

Genomic Map