Assay Details
Target Gene Details
Entrez Gene ID: | 22030 |
Gene Name: | TNF receptor-associated factor 2 |
Gene Aliases: |
AI325259 |
Location: |
Chr.2:25517982-25546940 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Traf2 | NM_001290413.1 | 11 | 2240 | NP_001277342.1 |
| NM_009422.3 | 11 | 2219 | NP_033448.2 | |
| AK052934.1 | 11 | 2168 | BAC35209.1 |
More Information
Set Membership: |
|

Genomic Map