Assay Details
Target Gene Details
Entrez Gene ID: | 622434 |
Gene Name: | Rho guanine nucleotide exchange factor (GEF) 26 |
Gene Aliases: |
4631416L12Rik, 8430436L14Rik, AI303572, AW491953, SGEF |
Location: |
Chr.3:62338777-62462221 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgef26 | NM_001081295.1 | 1 | 1492 | NP_001074764.1 |
| AK087060.1 | 1 | 1268 | ||
| AK144976.1 | 1 | 997 |
More Information
Set Membership: |
|

Genomic Map