Assay Details
Target Gene Details
Entrez Gene ID: | 66715 |
Gene Name: | HEN1 methyltransferase homolog 1 (Arabidopsis) |
Gene Aliases: |
4921515J06Rik, Hen1, mHEN1 |
Location: |
Chr.3:108938542-108960776 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Henmt1 | NM_025723.2 | 1 | 171 | NP_079999.2 |
| AK014900.1 | 1 | 141 | ||
| AK085587.1 | 1 | 171 | BAC39480.1 | |
| DV650789.1 | 1 | 238 |

Genomic Map