Assay Details
Target Gene Details
Entrez Gene ID: | 114249 |
Gene Name: | nephronectin |
Gene Aliases: |
1110009H02Rik, AA682063, AI314031, Nctn, POEM |
Location: |
Chr.3:132881745-132950291 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Npnt | NM_001029836.2 | 11 | 1721 | NP_001025007.1 |
| NM_001287101.1 | 13 | 1865 | NP_001274030.1 | |
| NM_001287102.1 | 12 | 1814 | NP_001274031.1 | |
| NM_001287103.1 | 11 | 1439 | NP_001274032.1 | |
| NM_033525.3 | 12 | 1772 | NP_277060.2 | |
| AB059656.1 | 11 | 1721 | BAB69692.1 | |
| AF397007.1 | 12 | 1566 | AAK84391.1 | |
| AF397008.1 | 13 | 1617 | AAK84392.1 | |
| AK050484.1 | 11 | 1625 | BAC34283.1 | |
| AK158764.1 | 11 | 1441 | BAE34649.1 | |
| AY035898.1 | 11 | 1596 | AAK96010.1 | |
| AY035899.1 | 12 | 1647 | AAK96011.1 | |
| BC046642.1 | 6 | 908 | AAH46642.1 | |
| BC068308.1 | 11 | 1590 | AAH68308.1 |

Genomic Map