Assay Details
Target Gene Details
Entrez Gene ID: | 67591 |
Gene Name: | ubiquitin-like 4B |
Gene Aliases: |
4930522D07Rik |
Location: |
Chr.3:107553698-107555073 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ubl4b | NM_026261.2 | 1 | 457 | NP_080537.1 |
| AK006408.1 | 1 | 383 | BAB24573.1 | |
| AK015868.1 | 1 | 458 | BAB30008.1 | |
| AK016746.1 | 1 | 414 | BAB30407.1 | |
| BC048443.1 | 1 | 396 | AAH48443.1 | |
| BC061071.1 | 1 | 422 | AAH61071.1 |

Genomic Map