Assay Details
Target Gene Details
Entrez Gene ID: | 73133 |
Gene Name: | RIKEN cDNA 3110021N24 gene |
Gene Aliases: |
2610002D03Rik |
Location: |
Chr.4:108719649-108781904 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 3110021N24Rik | NM_001254730.1 | 4 | 926 | NP_001241659.1 |
| AK011275.1 | 1 | 204 | ||
| AK014069.1 | 1 | 960 |
Target Gene Details
Entrez Gene ID: | 230597 |
Gene Name: | zinc finger, FYVE domain containing 9 |
Gene Aliases: |
E030027L17, Madhip, NSP, SARA, SMADIP |
Location: |
Chr.4:108637834-108781148 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfyve9 |

Genomic Map