Assay Details
Target Gene Details
Entrez Gene ID: | 20779 |
Gene Name: | Rous sarcoma oncogene |
Gene Aliases: |
AW259666, pp60c-src |
Location: |
Chr.2:157423901-157471838 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Src | NM_001025395.2 | 14 | 3517 | NP_001020566.1 |
| NM_009271.3 | 15 | 3535 | NP_033297.2 | |
| BC039953.1 | 14 | 3527 | AAH39953.1 |

Genomic Map