Assay Details
Target Gene Details
Entrez Gene ID: | 20811 |
Gene Name: | src-related kinase lacking C-terminal regulatory tyrosine and N-terminal myristylation sites |
Gene Aliases: |
A230069J08Rik, srm |
Location: |
Chr.2:181205562-181218783 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srms | NM_011481.3 | 8 | 2277 | NP_035611.3 |
| AK049582.1 | 8 | 1538 | BAC33825.1 | |
| BC120633.1 | ||||
| BC125325.1 | ||||
| D26186.1 | 8 | 2123 | BAA05331.1 | |
| D49427.1 | 8 | 2291 | BAA08406.1 |

Genomic Map