Assay Details
Target Gene Details
Entrez Gene ID: | 68911 |
Gene Name: | pygopus 2 |
Gene Aliases: |
1190004M21Rik |
Location: |
Chr.3:89430132-89435131 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pygo2 | NM_001293763.1 | 3 | 2527 | NP_001280692.1 |
| NM_001293766.1 | 2 | 2394 | NP_001280695.1 | |
| NM_001293767.1 | 3 | 2386 | NP_001280696.1 | |
| NM_001293768.1 | 3 | 2803 | NP_001280697.1 | |
| NM_026869.3 | 3 | 2469 | NP_081145.1 | |
| AK004486.1 | 1 | 1069 | BAE43180.1 | |
| AK132759.1 | 3 | 2470 | BAE21340.1 | |
| AK206147.1 | 1 | 239 | ||
| AY486147.1 | 3 | 2756 | AAS55435.1 | |
| BC025531.1 | 1 | 2220 | AAH25531.1 | |
| BC025640.1 | 3 | 2459 | AAH25640.1 |

Genomic Map