Assay Details
Target Gene Details
Entrez Gene ID: | 19338 |
Gene Name: | RAB33B, member RAS oncogene family |
Gene Aliases: |
- |
Location: |
Chr.3:51483966-51496228 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab33b | NM_016858.2 | 1 | 308 | NP_058554.1 |
| AB004665.1 | 1 | 258 | BAA22655.1 | |
| AK004974.1 | 1 | 44 | BAB23710.1 | |
| AK087967.1 | 1 | 48 | BAC40064.1 | |
| AK139821.1 | 1 | 293 | BAE24147.1 | |
| AK144706.1 | 1 | 45 | ||
| AK159674.1 | 1 | 43 | BAE35278.1 | |
| AK163783.1 | 1 | 295 | BAE37491.1 | |
| BC065076.1 | 1 | 308 | AAH65076.1 |

Genomic Map