Assay Details
Target Gene Details
Entrez Gene ID: | 100317 |
Gene Name: | expressed sequence AU040320 |
Gene Aliases: |
A730047D20Rik, Kiaa0319l |
Location: |
Chr.4:126753555-126869694 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AU040320 | NM_001035525.1 | NP_001030602.1 | ||
| NM_001035526.1 | NP_001030603.1 | |||
| NM_133886.2 | 21 | 4185 | NP_598647.1 | |
| AK043006.1 | BAC31432.1 | |||
| AK084668.1 | BAC39244.1 | |||
| AK161493.1 | 11 | 2248 | BAE36422.1 | |
| AK170261.1 | 21 | 4142 | BAE41669.1 | |
| AK206331.1 | 1 | 47 | ||
| AK211306.1 | 1 | 47 | ||
| BC022154.1 | 12 | 2597 | AAH22154.1 | |
| BC028869.1 | 21 | 4155 | AAH28869.1 |

Genomic Map